Learn More
Cole-Parmer Vacuum Regulator, 0 to 30" Hg, 2 cfm Max Capacity
Supplier:  Cole-Parmer 0706130
Encompass_Preferred
Order a vacuum regulator to monitor and control pressure levels. Each regulator comes with a gauge, except model, 98250-00.
Catalog No. NC9653660