Learn More
Fischer Technical Company Vacuum Regulator Kit - PILOT
SCI_ED
Catalog No. S2337145 Shop All Fischer Technical Company Products
Change view
Click to view available options
Flow Rate:
None
Catalog No. Flow Rate
S2337145 None
1 options
Catalog No. S2337145 Supplier Fischer Technical Company Supplier No. TLD3500K05
Add to Cart
Add to Cart
Vacuum Regulator Kit for the Pilot Vacuum Pumps (PILOT3000 and PILOT5000)
  • Regulator Kit includes: Dial Pressure Gauge, Hand Knob Control, 0.25 in. O.D. Hose barb, 1/8 in. NPT Exhaust Adapter, Built In Silencer.
  • Compatible with PILOT3000 and PILOT5000 Chemical Resistant Diaphragm Vacuum Pumps.
Certifications
SciEd
Specifications
Includes Dial Pressure Gauge, Hand Knob Control, 0.25 in. O.D. Hose barb, 1/8 in. NPT Exhaust Adapter, Built In Silencer.
Flow Rate None
Pump Type Diaphragm Vacuum Pump
Applications Vacuum Filtration
Maximum Pressure None
For Use With (Equipment) PILOT3000 and PILOT5000 Chemical Resistant Diaphragm Vacuum Pumps
Voltage 0/1 V
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov