Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Vacuum Seal
SDP

Catalog No. 02321108 Shop All Heidolph North America Products
Change view
Click to view available options
Includes:
FFKM Sealing Ring
FKM Sealing Ring
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
02-321-108 FFKM Sealing Ring
02-321-080 FKM Sealing Ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-321-108 Supplier Heidolph™ Supplier No. 036210033
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The highly durable and long lasting PTFE vacuum seals with an FKM or an FFKM sealing ring to achieve maximum tightness and save on long-term spare parts costs.

  • PTFE spare seal
  • Choose between an FKM or an FFKM sealing ring
  • Materials offer very good chemical resistance and the high quality sealing properties
  • Long running time without abrasion
  • Highest density
  • FDA compliant

Specifications

Color Natural
Material PTFE
Type Vacuum Seal
For Use With (Equipment) Evaporators
Includes FFKM Sealing Ring
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.