Learn More
Chemglass Life Sciences Airfree Portable Vacuum System, Complete To Include:
SDP
Catalog No. AF030001
Encompass_Preferred
Airfree Portable Vacuum System, Complete. The Chemglass Airfree portable vacuum system has been designed to be a totally self-contained system for use in vacuum distillations, sample degassing, vapor pressure and other preparative work requiring vacuum to 1 micron. Please Note: System is supplied complete as shown with the exception of the Dewar flask, lab jack and port adapters; all of which are available as options.Complete system is shipped partially assembled with instructions in a wooden crate via common carrier, FOB Vineland, NJ. System can be customized with any of the listed optional items, or components can be purchased individually using the part numbers on the following pages. Approx. dimensions are 28 W x 20 D x 70 H.
Catalog No. AF030001 Supplier Chemglass Life Sciences Supplier No. AF030001
Add to Cart
Add to Cart
Request a Quote
Airfree Portable Vacuum System, Complete. The Chemglass Airfree portable vacuum system has been designed to be a totally self-contained system for use in vacuum distillations, sample degassing, vapor pressure and other preparative work requiring vacuum to 1 micron. Please Note: System is supplied complete as shown with the exception of the Dewar flask, lab jack and port adapters; all of which are available as options.Complete system is shipped partially assembled with instructions in a wooden crate via common carrier, FOB Vineland, NJ. System can be customized with any of the listed optional items, or components can be purchased individually using the part numbers on the following pages. Approx. dimensions are 28 W x 20 D x 70 H.