Learn More
Chemglass Life Sciences Vacuum System, Self-Adjusting, PC3002 Vario Select, Touch Screen Controller, 100-120V
SDP
Catalog No. 501979516
Encompass_Preferred
Vacuum System, Self-Adjusting, PC3002 Vario Select, Touch Screen Controller, 100-120V. The PC3002 VARIO select is the best choice vacuum system for bench-top sized rotary evaporators and other evaporative applications. The PC3002 VARIO select controls the vacuum in your rotary evaporator with minimal supervision, continuously optimizing to changing vapor conditions without programming. Just press "start" and the PC3002 VARIO select provides faster evaporation and tighter control, while freeing you up to do more important work. It features a VARIO version of our MD1C vacuum pump, with integrated solvent recovery in a compact, attractive package.Touch Screen Controller:The integrated VACUU-SELECT controller comes with automatic operating modes for most common lab vacuum applications. Drag and drop menus let you set up your own custom operations in minutes.
Catalog No. 50-197-9516 Supplier Chemglass Life Sciences Supplier No. CG4802102
Add to Cart
Add to Cart
Request a Quote
Vacuum System, Self-Adjusting, PC3002 Vario Select, Touch Screen Controller, 100-120V. The PC3002 VARIO select is the best choice vacuum system for bench-top sized rotary evaporators and other evaporative applications. The PC3002 VARIO select controls the vacuum in your rotary evaporator with minimal supervision, continuously optimizing to changing vapor conditions without programming. Just press "start" and the PC3002 VARIO select provides faster evaporation and tighter control, while freeing you up to do more important work. It features a VARIO version of our MD1C vacuum pump, with integrated solvent recovery in a compact, attractive package.Touch Screen Controller:The integrated VACUU-SELECT controller comes with automatic operating modes for most common lab vacuum applications. Drag and drop menus let you set up your own custom operations in minutes.