Learn More
Chemglass Life Sciences Vacuum Trap, 35mm Body OD x 250mm Body Length, 16mm OD Side and Inner Tubes
SDP
Catalog No. CG451203
Encompass_Preferred
Vacuum Trap, 35mm Body OD x 250mm Body Length, 16mm OD Side and Inner Tubes. Similar to CG-4510 but with top inlet tube bent at a 90º angle.
Catalog No. CG451203 Supplier Chemglass Life Sciences Supplier No. CG451203
Vacuum Trap, 35mm Body OD x 250mm Body Length, 16mm OD Side and Inner Tubes. Similar to CG-4510 but with top inlet tube bent at a 90º angle.