Learn More
Chemglass Life Sciences Vacuum Trap, Complete, 60mm Body OD x 300mm Overall Length, 30mm OD Side and Inner Tubes, 55/50 Jt.
SDP
Catalog No. CG451614
Encompass_Preferred
Vacuum Trap, Dual Chamber, Complete. Dual chamber vacuum trap affords increased condensing capacity in a much shorter length. The dry ice cold fingers have 55/50 inner joints that fit into the outer joints on the main trap body and have a capacity of approx. 300mL each. Lower stopcock is a PTFE 4mm ¿T¿-bore which permits draining condensate chambers one at a time. Serrated connections at the top of the condensate chambers have an O.D. of 10mm. Complete trap assembly is approx. 545mm in height x 220mm wide.
Catalog No. CG451614 Supplier Chemglass Life Sciences Supplier No. CG451614
Vacuum Trap, Dual Chamber, Complete. Dual chamber vacuum trap affords increased condensing capacity in a much shorter length. The dry ice cold fingers have 55/50 inner joints that fit into the outer joints on the main trap body and have a capacity of approx. 300mL each. Lower stopcock is a PTFE 4mm ¿T¿-bore which permits draining condensate chambers one at a time. Serrated connections at the top of the condensate chambers have an O.D. of 10mm. Complete trap assembly is approx. 545mm in height x 220mm wide.