Learn More
Chemglass Life Sciences Vacuum Trap, 500mL Capacity, Fits 1900mL Dewar Flask, 360mm OAH x 105mm Width, Complete
SDP
Catalog No. CG453101
Encompass_Preferred
Vacuum Trap, 500mL Capacity, Fits 1900mL Dewar Flask, 360mm OAH x 130mm Width, Complete. Similar to CG-4529, but having a 40mm o-ring joint to connect the trap body to the stopper. O-ring joint provides for a vacuum-tight grease-free seal and permits the trap body to be easily disconnected from the stopper for cleaning. Complete trap is supplied with a stopper, trap body and CG-150-09 clamp.
Catalog No. CG453101 Supplier Chemglass Life Sciences Supplier No. CG453101
Vacuum Trap, 500mL Capacity, Fits 1900mL Dewar Flask, 360mm OAH x 130mm Width, Complete. Similar to CG-4529, but having a 40mm o-ring joint to connect the trap body to the stopper. O-ring joint provides for a vacuum-tight grease-free seal and permits the trap body to be easily disconnected from the stopper for cleaning. Complete trap is supplied with a stopper, trap body and CG-150-09 clamp.