Learn More
Chemglass Life Sciences Vacuum Trap, Complete
SDP
Supplier:  Chemglass Life Sciences CG453001
Encompass_Preferred
Vacuum Trap, Complete. Trap is designed for high volume solvent removal by vacuum. The unique design of the stopper decreases the chance of freezing by allowing vapor to pass through the holes in the side of the inner tube. The 55/50 standard taper joint connects the trap body with the stopper. Hose connections are 15mm OD at the largest serration.
Catalog No. CG453001