Learn More
Chemglass Life Sciences Vacuum Trap, Complete, Trap Bottle 60mm OD x 300mm Overall Length, 30mm OD Side and Inner Tubes, 55/50 Jt.
SDP
Catalog No. CG451414
Encompass_Preferred
Vacuum Trap, Complete, Trap Bottle 60mm OD x 300mm Overall Length, 30mm OD Side and Inner Tubes, 55/50 Jt. Two piece vacuum trap connected with an inner and outer joint to facilitate cleaning.
Catalog No. CG451414 Supplier Chemglass Life Sciences Supplier No. CG451414
Vacuum Trap, Complete, Trap Bottle 60mm OD x 300mm Overall Length, 30mm OD Side and Inner Tubes, 55/50 Jt. Two piece vacuum trap connected with an inner and outer joint to facilitate cleaning.