Learn More
Chemglass Life Sciences Vacuum Trap, 1000mL Capacity, Fits 4300mL Dewar Flask, 340mm OAH x 130mm Width, Complete
SDP
Catalog No. CG452902
Encompass_Preferred
Vacuum Trap, 1000mL Capacity, Fits 4300mL Dewar Flask, 340mm OAH x 130mm Width, Complete. Trap is designed for large volume solvent removal by vacuum. The 40/35 standard taper joint connects trap body with stopper. Connections are 1/2in O.D. Trap is designed to fit into standard CG-1593 through CG-1595 Dewar flasks.
Catalog No. CG452902 Supplier Chemglass Life Sciences Supplier No. CG452902
Vacuum Trap, 1000mL Capacity, Fits 4300mL Dewar Flask, 340mm OAH x 130mm Width, Complete. Trap is designed for large volume solvent removal by vacuum. The 40/35 standard taper joint connects trap body with stopper. Connections are 1/2in O.D. Trap is designed to fit into standard CG-1593 through CG-1595 Dewar flasks.