Learn More
Synthware™ Vacuum Trap - Ball Joints
GREENER_CHOICESDP
Catalog No. 31501802 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
45/50
Catalog No. Standard Taper Joint
31-501-802 45/50
1 options
Catalog No. 31-501-802 Supplier Synthware™ Supplier No. V415045
Add to Cart
Add to Cart
For use with vacuum manifolds having 35/20 socket joints.
Specifications
For Use With (Application) Organic Synthesis
Type 35/20 Ball Joints
Material Borosilicate Glass
For Use With (Equipment) Vacuum manifolds having 35/20 socket joints
Quantity 1 Ea.
Standard Taper Joint 45/50