Learn More
Chemglass Life Sciences Vacuum Trap, Complete, Trap Bottle 38mm OD x 225mm Overall Length, 13mm OD Side and Inner Tubes, 34/45 Jt.
SDP
Catalog No. CG451404
Encompass_Preferred
Vacuum Trap, Complete, Trap Bottle 38mm OD x 225mm Overall Length, 13mm OD Side and Inner Tubes, 34/45 Jt. Two piece vacuum trap connected with an inner and outer joint to facilitate cleaning.
Catalog No. CG451404 Supplier Chemglass Life Sciences Supplier No. CG451404
Vacuum Trap, Complete, Trap Bottle 38mm OD x 225mm Overall Length, 13mm OD Side and Inner Tubes, 34/45 Jt. Two piece vacuum trap connected with an inner and outer joint to facilitate cleaning.