Learn More
Chemglass Life Sciences Vacuum Trap, O-Ring, Airfree, Schlenk, Complete
SDP
Supplier:  Chemglass Life Sciences AF020001
Encompass_Preferred
Vacuum Trap, O-Ring, Airfree, Schlenk, Complete. Two-piece vacuum trap with #15 o-ring joints on both the inlet and outlet ports. The seal between the trap stopper and body is made with a 57mm flat o-ring flange for total grease-free operation when used with either the AF-0060 or AF-0061 manifolds. Trap body is 57mm OD x 300mm in length to be used with a 4,300mL Dewar flask (CG-1593-05 or CG-1594-05). Complete item consists of the trap stopper and body, one CG-139-02 horseshoe clamp, one #331 Viton o-ring and two #116 Viton o-rings.
Catalog No. AF020001