Learn More
Chemglass Life Sciences Vacuum Trap, Complete, 54mm Body OD x 275mm Overall Length, 25mm OD Side and Inner Tubes, 50/50 Jt.
SDP
Catalog No. CG451610
Encompass_Preferred
Vacuum Trap, Complete, 54mm Body OD x 275mm Overall Length, 25mm OD Side and Inner Tubes, 50/50 Jt. Similar to CG-4514, but with the top inlet tube bent at a 90º angle.
Catalog No. CG451610 Supplier Chemglass Life Sciences Supplier No. CG451610
Vacuum Trap, Complete, 54mm Body OD x 275mm Overall Length, 25mm OD Side and Inner Tubes, 50/50 Jt. Similar to CG-4514, but with the top inlet tube bent at a 90º angle.