Learn More
Chemglass Life Sciences Vacuum Trap, Complete, 45mm Body OD x 250mm Overall Length, 16mm OD Side and Inner Tubes, 40/50 Jt.
SDP
Catalog No. CG451607
Encompass_Preferred
Vacuum Trap, Complete, 45mm Body OD x 250mm Overall Length, 16mm OD Side and Inner Tubes, 40/50 Jt. Similar to CG-4514, but with the top inlet tube bent at a 90º angle.
Catalog No. CG451607 Supplier Chemglass Life Sciences Supplier No. CG451607
Vacuum Trap, Complete, 45mm Body OD x 250mm Overall Length, 16mm OD Side and Inner Tubes, 40/50 Jt. Similar to CG-4514, but with the top inlet tube bent at a 90º angle.