Learn More
Ace Glass, Inc. Vacuum trap, dry ice, #50 Ace-Thred top
SDP
Catalog No. 50976332
Encompass_Preferred
Dry ice finger with #50 Ace-Thred top, for twin-chambered dry ice condenser- vacuum traps. For use with Rotary Evaporators. Fingers are secured in main body via the #50 Ace-Thred, bushings, and o-rings, allowing for easy cleaning.
Catalog No. 50-976-332 Supplier Ace Glass, Inc. Supplier No. 874804
Dry ice finger with #50 Ace-Thred top, for twin-chambered dry ice condenser- vacuum traps. For use with Rotary Evaporators. Fingers are secured in main body via the #50 Ace-Thred, bushings, and o-rings, allowing for easy cleaning.