Learn More
Chemglass Life Sciences Vacuum Trap STOPPER ONLY, Airfree, Schlenk, 57mm O-ring Flange
SDP
Catalog No. AF020003
Encompass_Preferred
Vacuum Trap STOPPER ONLY, Airfree, Schlenk, 57mm O-ring Flange. The seal between the trap stopper and body is made with a 57mm flat o-ring flange for total grease-free operation when used with either the AF-0060 or AF-0061 manifolds. Complete item consists of the trap stopper and body, one CG-139-02 horseshoe clamp, one #331 Viton o-ring and two #116 Viton o-rings.
Catalog No. AF020003 Supplier Chemglass Life Sciences Supplier No. AF020003
Vacuum Trap STOPPER ONLY, Airfree, Schlenk, 57mm O-ring Flange. The seal between the trap stopper and body is made with a 57mm flat o-ring flange for total grease-free operation when used with either the AF-0060 or AF-0061 manifolds. Complete item consists of the trap stopper and body, one CG-139-02 horseshoe clamp, one #331 Viton o-ring and two #116 Viton o-rings.