Learn More
Chemglass Life Sciences Vacuum Trap, O-Ring Style, Lower Body Only, #50 O-Ring Joint
SDP
Catalog No. 501531740
Encompass_Preferred
Vacuum Trap, O-Ring Style, Lower Body Only, #50 O-Ring Joint. Similar to CG-4515 traps but having an o-ring joint to connect the trap body to the stopper. Serrated hose connections are 13.5mm at the smallest serration and 15.5mm at the largest serration. LOWER BODY ONLY! Pictured is the complete vacuum trap assembly.
Catalog No. 50-153-1740 Supplier Chemglass Life Sciences Supplier No. CG451322
Vacuum Trap, O-Ring Style, Lower Body Only, #50 O-Ring Joint. Similar to CG-4515 traps but having an o-ring joint to connect the trap body to the stopper. Serrated hose connections are 13.5mm at the smallest serration and 15.5mm at the largest serration. LOWER BODY ONLY! Pictured is the complete vacuum trap assembly.