Learn More
Chemglass Life Sciences Vacuum Trap, O-Ring Style, Complete, #40 O-Ring Joint, 45mm OD x 250mm Length
SDP
Catalog No. 50873447
Encompass_Preferred
Vacuum Trap, O-Ring Style, Complete, #40 O-Ring Joint, 45mm OD x 250mm Length. Similar to CG-4515 traps but having an o-ring joint to connect the trap body to the stopper. O-ring joint provides for a vacuum tight, grease free seal and permits the trap body to be easily disconnected from the stopper for cleaning. Serrated hose connections are 13.5mm at the smallest serration and 15.5mm at the largest serration. Complete trap is supplied with a stopper, lower body, viton o-ring, and Stainless Steel pinch clamp.
Catalog No. 50-873-447 Supplier Chemglass Life Sciences Supplier No. CG451311
Vacuum Trap, O-Ring Style, Complete, #40 O-Ring Joint, 45mm OD x 250mm Length. Similar to CG-4515 traps but having an o-ring joint to connect the trap body to the stopper. O-ring joint provides for a vacuum tight, grease free seal and permits the trap body to be easily disconnected from the stopper for cleaning. Serrated hose connections are 13.5mm at the smallest serration and 15.5mm at the largest serration. Complete trap is supplied with a stopper, lower body, viton o-ring, and Stainless Steel pinch clamp.