Learn More
Foxx Life Sciences Vacuum Trap System, Safety Coated Glass Bottles 10L+2L
Catalog No. 5000705950
Encompass_Preferred
Vactrap™ Rhino Glass Bottle vacuum aspirator collection system is designed to meet strict CDC laboratory guidelines for handling biohazardous waste. This Fluid Aspiration System utilizes shatter-resistant, safety coated glass bottles that offer superior chemical stability & liquid containment in the event of breakage.
Features
Vacuum Trap Assembly with coated glass bottle for shatter resistance & liquid containment.
40 CFR 264.175 compliant aspirator system has a compact, tip-resistant secondary container for stability.
Bottles made with BOROSILICATE 3.3 are chemically inert & ideal for harsh chemicals.
Standard Vacuum Trap with GL45 VersaCap closure system with rotating adapter prevents tube tangling.
MADE IN USA
Applications
Aspiration of cell culture media
Vacuum pump trap kit
Waste collection System for high-purity processes
Ideal for house vacuum or vacuum pump trap systems
Aspirator Bottle Assembly
Catalog No. 50-007-05950 Supplier Foxx Life Sciences Supplier No. 30RA203FLS
Vactrap™ Rhino Glass Bottle vacuum aspirator collection system is designed to meet strict CDC laboratory guidelines for handling biohazardous waste. This Fluid Aspiration System utilizes shatter-resistant, safety coated glass bottles that offer superior chemical stability & liquid containment in the event of breakage.
Features
Vacuum Trap Assembly with coated glass bottle for shatter resistance & liquid containment.
40 CFR 264.175 compliant aspirator system has a compact, tip-resistant secondary container for stability.
Bottles made with BOROSILICATE 3.3 are chemically inert & ideal for harsh chemicals.
Standard Vacuum Trap with GL45 VersaCap closure system with rotating adapter prevents tube tangling.
MADE IN USA
Applications
Aspiration of cell culture media
Vacuum pump trap kit
Waste collection System for high-purity processes
Ideal for house vacuum or vacuum pump trap systems
Aspirator Bottle Assembly