Learn More
AVANTOR MASTERFLEX (VWR LLC) Black Rubber Vacuum Tubing, 1/4"" ID x 5/8"" OD, 25ft. 96143-06
Catalog No. NC9568608
Encompass
Applications: Vacuum and pressure applications for general lab and industrial use Characteristics: Thick wall, flexible, opaque Temperature Range: 0¿F to 150¿F Sterilize: Ethylene oxide or autoclave for 30 minutes at 15 psi
Catalog No. NC9568608 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9614306
Applications: Vacuum and pressure applications for general lab and industrial use Characteristics: Thick wall, flexible, opaque Temperature Range: 0¿F to 150¿F Sterilize: Ethylene oxide or autoclave for 30 minutes at 15 psi