Learn More
Heidolph™ Vacuum Valve
SDP
Catalog No. 036308071 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Hei-VAP Expert Control and Hei-VAP Ultimate Control Rotary Evaporator
Catalog No. For Use With (Equipment)
03-630-8071 Hei-VAP Expert Control and Hei-VAP Ultimate Control Rotary Evaporator
1 options
Catalog No. 03-630-8071 Supplier Heidolph™ Supplier No. 036308071
Add to Cart
Add to Cart
Required for Hei-VAP Control models with valve-controlled vacuum pump. Can be positioned for easy access and cleaning.
Not required for manual vacuum control
Specifications
Type Rotary Evaporator Accessory
For Use With (Equipment) Hei-VAP Expert Control and Hei-VAP Ultimate Control Rotary Evaporator
Includes Valve
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov