Learn More
Sino Biological Vaccinia Virus B18R / B19R Protein (His Tag)
Supplier:  Sino Biological 40020V08B100
Encompass_Preferred
A DNA sequence encoding the Vaccinia Virus B18R (P25213) (Met1-Glu351) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-255-3751