Learn More
Medchemexpress LLC HY-101277 50mg , Vadadustat CAS:1000025-07-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10127750MG
Encompass_Preferred
HY-101277 50mg Vadadustat CAS: 1000025-07-9 Vadadustat (PG-1016548) is a titratable, oral hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor. Vadadustat is an erythropoiesis-stimulating agent and has the potential for anemia treatment in chronic kidney disease in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9881