Learn More
Medchemexpress LLC HY-10964 10mg , Vadimezan CAS:117570-53-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1096410MG
Encompass_Preferred
HY-10964 10mg Vadimezan CAS: 117570-53-3 Vadimezan (DMXAA; ASA-404), the tumor vascular disrupting agent (tumor-VDA), is a murine agonist of the stimulator of interferon genes (STING) and also a potent inducer of type I IFNs and other cytokines. Vadimezan has anti-influenza virus H1N1-PR8 activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9126