Learn More
ATCC VA-ES-BJ; Epithelioid Sarcoma; Human (Homo sapiens)
Catalog No. 502384912
Encompass
VA-ES-BJ is a cell line exhibiting epithelial morphology that was isolated from a 41-day-old, White male patient with epithelioid sarcoma. It has applications in immunology and cancer research.
Catalog No. 50-238-4912 Supplier ATCC Supplier No. CRL2138
VA-ES-BJ is a cell line exhibiting epithelial morphology that was isolated from a 41-day-old, White male patient with epithelioid sarcoma. It has applications in immunology and cancer research.