Learn More
Medchemexpress LLC Vafidemstat | 1357362-02-7 | 98.0% | 336.39 | 250 MG
SDP
Catalog No. 5000439692
Encompass_Preferred
Vafidemstat (ORY-2001) is an oral, brain penetrant, dual lysine-specific histone demethylase (LSD1)/MAO-B inhibitor. It is a novel epigenetic agent developed for the treatment of neurodegenerative diseases. It modulates LSD1, which participates in transcription regulation complexes, to adjust transcriptional imbalances in neurodegenerative diseases and to address neuroinflammation and cognitive deficits. Vafidemstat (ORY-2001) can also be used in the treatment of Alzheimer's disease.
  • Oral, brain penetrant.
  • Dual lysine-specific histone demethylase (LSD1)/MAO-B inhibitor.
  • Novel epigenetic agent for the treatment of neurodegenerative diseases.
  • Modulates LSD1 to tweak transcriptional imbalances in neurodegenerative disease.
  • Redresses neuroinflammation and cognitive deficit.
  • Can be used in the treatment of Alzheimer's disease.
Catalog No. 50-004-39692 Supplier Medchemexpress LLC Supplier No. 250MGHY112623
Vafidemstat (ORY-2001) is an oral, brain penetrant, dual lysine-specific histone demethylase (LSD1)/MAO-B inhibitor. It is a novel epigenetic agent developed for the treatment of neurodegenerative diseases. It modulates LSD1, which participates in transcription regulation complexes, to adjust transcriptional imbalances in neurodegenerative diseases and to address neuroinflammation and cognitive deficits. Vafidemstat (ORY-2001) can also be used in the treatment of Alzheimer's disease.
  • Oral, brain penetrant.
  • Dual lysine-specific histone demethylase (LSD1)/MAO-B inhibitor.
  • Novel epigenetic agent for the treatment of neurodegenerative diseases.
  • Modulates LSD1 to tweak transcriptional imbalances in neurodegenerative disease.
  • Redresses neuroinflammation and cognitive deficit.
  • Can be used in the treatment of Alzheimer's disease.