Learn More
ATCC Vaginal Microbiome; Whole Cell Mix
Catalog No. 502382321
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product is prepared as an even mixture of whole cells and comprises fully sequenced, characterized, and authenticated bacterial species observed in normal and atypical vaginal microbial communities. Each order for this product is provided with access to run 10 analyses on One Codex, the leading bioinformatics platform for microbial genomics and metagenomics. Specification Range: 1.2x108 cells/vial ± 1 log* *Whole cell concentrations indicate ATCC manufacturing specifications and are provided as a reference only
Catalog No. 50-238-2321 Supplier ATCC Supplier No. MSA2007
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product is prepared as an even mixture of whole cells and comprises fully sequenced, characterized, and authenticated bacterial species observed in normal and atypical vaginal microbial communities. Each order for this product is provided with access to run 10 analyses on One Codex, the leading bioinformatics platform for microbial genomics and metagenomics. Specification Range: 1.2x108 cells/vial ± 1 log* *Whole cell concentrations indicate ATCC manufacturing specifications and are provided as a reference only