Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Vaginal Verification Panel
SDP

Catalog No. 23046873 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
120 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-873 120 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-873 Supplier Microbiologics™ Supplier No. 8208
Add to Cart
Add to Cart
This item is not returnable. View return policy

Microbiologics™ Vaginal Verification Panel includes 2 boxes containing 20 individually packaged inactivated microorganism pellets for in vitro diagnostic use.

  • Pellets are stable at room-temperature
  • For in vitro diagnostic use
  • Includes 2 boxes containing 20 individually packaged inactivated microorganism pellets of each pool listed below (120 pellets/vials total):
    • Pool 1 (20 pellets/vials)
      Lactobacillus crispatus derived from NCIMB 702752
    • Pool 2 (20 pellets/vials)
      Gardnerella vaginalis derived from NCTC 10915
    • Pool 3 (20 pellets/vials)
      Atopobium vaginae derived from NCTC 13935
      Recombinant BVAB2
      Candida albicans derived from NCPF 3179
      Gardnerella vaginalis derived from NCTC 10915
    • Pool 4 (20 pellets/vials)
      Atopobium vaginae derived from NCTC 13935
      Recombinant BVAB2
      Gardnerella vaginalis derived from NCTC 10915
      Trichomonas vaginalis Strain GL0028
    • Pool 5 (20 pellets/vials)
      Atopobium vaginae derived from NCTC 13935
      Candida glabrata derived from NCPF 8018
      Gardnerella vaginalis derived from NCTC 10915
      Lactobacillus crispatus derived from NCIMB 702752
    • Pool 6 (20 pellets/vials)
      Atopobium vaginae derived from NCTC 13935
      Candida krusei derived from NCPF 3100
      Gardnerella vaginalis derived from NCTC 10915
      Lactobacillus crispatus derived from NCIMB 702752

Specifications

Certifications/Compliance IVD, CE Marked
Content And Storage Room Temperature
Format Helix Elite
Manufacturing Origin United States
Product Type Helix Elite Control Reference Panels and Standards
For Use With (Application) For in vitro diagnostic use
Packaging Type Plastic Box
Quantity 120 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.