Learn More
Med Vet International Bovine Artificial Vagina Kit - 14", J0125B, Each
Supplier:  Med Vet International JOR125B
Encompass_Preferred
Bovine Artificial Vagina Kit - 14". Black rubber mold tube with flanges on ends. Features: • 12" heavyweight latex liner • Lightweight latex end collection cone • 2 securing wide rubber bands • 15ml centrifuge tube
Catalog No. 50-004-95692