Learn More
Instech Mating tether for Vascular Access Harness™ or Button for mice. 7in (18cm), polyurethane in fluid path. Compatible with Instech 25ga plastic swivels. For mouse infusion and blood sampling.
SDP
Catalog No. 5081243
Encompass
The Instech VAH62T is a sterile mouse tether designed for use with the Vascular Access Harness™ (VAH62AB series), providing a secure, low‑dead‑volume connection for infusion or blood sampling. Built to mate with 25ga low‑torque swivels, the 7in (18cm) spring tether protects the externalized catheter while allowing free movement during studies, ensuring smooth, aseptic connection to the harness port.
Features
  • Sterile tether for mouse Vascular Access Harness™ systems
  • 7in (18cm) spring tether length
  • Compatible with 25ga low‑torque swivels
  • Provides clean, low‑volume connection to harness port
  • Supports mouse infusion and blood sampling workflows
  • Protects externalized catheter while allowing mobility
Catalog No. 50-812-43 Supplier Instech Supplier No. VAH62T
The Instech VAH62T is a sterile mouse tether designed for use with the Vascular Access Harness™ (VAH62AB series), providing a secure, low‑dead‑volume connection for infusion or blood sampling. Built to mate with 25ga low‑torque swivels, the 7in (18cm) spring tether protects the externalized catheter while allowing free movement during studies, ensuring smooth, aseptic connection to the harness port.
Features
  • Sterile tether for mouse Vascular Access Harness™ systems
  • 7in (18cm) spring tether length
  • Compatible with 25ga low‑torque swivels
  • Provides clean, low‑volume connection to harness port
  • Supports mouse infusion and blood sampling workflows
  • Protects externalized catheter while allowing mobility