Learn More
Instech Mating tether for Vascular Access Harness™ for rats. 12in (30cm), polyurethane in fluid path. Compatible with Instech swivels. For rat infusion and blood sampling. See part no KVAH95T for a kit including swivel and tubing to pump.
SDP
Catalog No. 5081247
Encompass
The Instech VAH95T is a sterile rat Vascular Access Harness tether assembly designed for continuous infusion or blood sampling, featuring 024 polyurethane tubing in the fluid path for durability and low dead volume. Measuring 12in (30cm) in length, it serves as the mating tether for Vascular Access Harness™ systems for rats and is fully compatible with Instech swivels for smooth rotational movement during tethered studies.
Features
  • Sterile rat VAH tether assembly
  • 12in (30cm) length for flexible routing
  • 024 PU tubing for a reliable low‑dead‑volume fluid path
  • Mating tether for Vascular Access Harness™ for rats
  • Compatible with Instech low‑torque swivel systems
  • Suitable for rat infusion and blood sampling
  • See part no. KVAH95T for kit with swivel and pump‑line tubing
Catalog No. 50-812-47 Supplier Instech Supplier No. VAH95T
The Instech VAH95T is a sterile rat Vascular Access Harness tether assembly designed for continuous infusion or blood sampling, featuring 024 polyurethane tubing in the fluid path for durability and low dead volume. Measuring 12in (30cm) in length, it serves as the mating tether for Vascular Access Harness™ systems for rats and is fully compatible with Instech swivels for smooth rotational movement during tethered studies.
Features
  • Sterile rat VAH tether assembly
  • 12in (30cm) length for flexible routing
  • 024 PU tubing for a reliable low‑dead‑volume fluid path
  • Mating tether for Vascular Access Harness™ for rats
  • Compatible with Instech low‑torque swivel systems
  • Suitable for rat infusion and blood sampling
  • See part no. KVAH95T for kit with swivel and pump‑line tubing