Learn More
Instech Mating tether for dual channel rat Vascular Access Harness; only one tube connected. 12in. Sterile. Mates with part no VAHD115AB. For bile sampling.
SDP
Catalog No. 5081259
Encompass
The Instech VAHD115T1 is a sterile one‑channel tether assembly designed for use with dual channel VAH rat harnesses, providing a single active line connected to channel A for bile sampling workflows. Measuring 12in in length, it mates directly with the VAHD115AB harness and offers a simple, low‑dead‑volume connection optimized for collecting bile without volume replacement through the second channel.
Features
  • Mating tether for dual channel VAH rat harnesses
  • One active tube connected to channel A
  • 12in tether length for flexible setup
  • Sterile assembly for bile sampling procedures
  • Designed for use with VAHD115AB
Catalog No. 50-812-59 Supplier Instech Supplier No. VAHD115T1
The Instech VAHD115T1 is a sterile one‑channel tether assembly designed for use with dual channel VAH rat harnesses, providing a single active line connected to channel A for bile sampling workflows. Measuring 12in in length, it mates directly with the VAHD115AB harness and offers a simple, low‑dead‑volume connection optimized for collecting bile without volume replacement through the second channel.
Features
  • Mating tether for dual channel VAH rat harnesses
  • One active tube connected to channel A
  • 12in tether length for flexible setup
  • Sterile assembly for bile sampling procedures
  • Designed for use with VAHD115AB