Learn More
Instech Two channel Vascular Access Harness tether assembly, 12in, sterile
SDP
Catalog No. 5081257
Encompass
The two-channel Vascular Access Harness (VAH) connects two independent channels as simply as the standard VAH connects one. Install the harness when the catheters are implanted or connect after recovery.
Access directly with a syringe fitted with a VAH6M injector or plug in a mating tether for continuous access.
Mates with part no VAHD115AB (Two channel rat VAH harness, injector, sterile)
This item is not returnable. View return policy
Catalog No. 50-812-57 Supplier Instech Supplier No. VAHD115T
The two-channel Vascular Access Harness (VAH) connects two independent channels as simply as the standard VAH connects one. Install the harness when the catheters are implanted or connect after recovery.
Access directly with a syringe fitted with a VAH6M injector or plug in a mating tether for continuous access.
Mates with part no VAHD115AB (Two channel rat VAH harness, injector, sterile)