Learn More
VAZYME INTERNATIONAL LLC Vazyme International VAHTS DNA Clean Beads, 5 mL, DNA Purification & NGS Library Prep
Catalog No. NC3979769
Encompass
VAHTS DNA Clean Beads utilize SPRI (Solid Phase Reverse Immobilization) technology for efficient DNA purification and fragment size selection. They are designed for next-generation sequencing (NGS) library preparation and are compatible with various DNA/RNA library construction protocols. These beads provide a cost-effective alternative while offering comparable performance to other widely used products, ensuring consistent yield and size distribution of libraries.
  • Based on SPRI technology for DNA purification and fragment size selection.
  • Suitable for next-generation sequencing (NGS) library preparation.
  • Compatible with most DNA or RNA library construction protocols.
  • Offers consistent yield and size distribution of libraries.
  • All components can be stored at 2°C to 8°C for one year.
Catalog No. NC3979769 Supplier VAZYME INTERNATIONAL LLC Supplier No. N41101
VAHTS DNA Clean Beads utilize SPRI (Solid Phase Reverse Immobilization) technology for efficient DNA purification and fragment size selection. They are designed for next-generation sequencing (NGS) library preparation and are compatible with various DNA/RNA library construction protocols. These beads provide a cost-effective alternative while offering comparable performance to other widely used products, ensuring consistent yield and size distribution of libraries.
  • Based on SPRI technology for DNA purification and fragment size selection.
  • Suitable for next-generation sequencing (NGS) library preparation.
  • Compatible with most DNA or RNA library construction protocols.
  • Offers consistent yield and size distribution of libraries.
  • All components can be stored at 2°C to 8°C for one year.