Learn More
VAI Alcoh-Glove Nonlinting Wipers
Catalog No. 19546787 Shop All VAI Products
Change view
Click to view available options
Saturation Solution:
70% IPA, 30% Deionised Water
Catalog No. Saturation Solution
19-546-787 70% IPA, 30% Deionised Water
1 options
This item is not returnable. View return policy
Catalog No. 19-546-787 Supplier VAI Supplier No. AG0470%9053
Add to Cart
Add to Cart
This item is not returnable. View return policy
Alcoh-Glove Nonlinting Wipers provide 100% coverage of the hand. It is a non-linting, non-shedding polyester tube that is sewn at one end. The glove is saturated with 70% USP Isopropyl Alcohol and 30% USP WFI, individually packaged, and terminally sterilized.
  • Laundered in ISO 4 cleanroom conditions (Grade A/B, Former Class 10)
  • Filtered at 0.2 microns
  • Individually packed and folded for easy removal
  • Completely lot traceable
  • Tested according to current USP Compendium
  • Individually labeled with lot number and expiration date
  • Gamma irradiated at 10-6 SAL
Specifications
Product Type Glove Wipe
Material Polyester
Edges Contoured
For Use With (Application) Designed for sterile operations that demand the use of an individually packaged sterile saturated wipe
Saturation Solution 70% IPA, 30% Deionised Water
Sterility Sterile