Learn More
VAI Core2Clean System™ Mop Frame
Catalog No. 19546796 Shop All VAI Products
Change view
Click to view available options
Length (English):
7 in.
Catalog No. Length (English)
19-546-796 7 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-546-796 Supplier VAI Supplier No. C2C10027
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
The Core2Clean System™ Mop Frame is designed to address the application of cleaning and disinfecting agents to the surface in a meaningful and manageable methodology.
  • Made of stainless steel and can be steam sterilized to eliminate the concerns for introduction of viable contamination to the controlled environment from the components themselves.
  • Eliminate concerns for changing of dirtied cleaning solution
  • Incorporate a trigger activated dispensing mop, a sprayer and a fogger all in one unit
  • Present a sufficient wetting of the surface unavailable from other mopping sytems
  • Can reduce cleaning time by 50% and is easily moved from area to area by a 4-wheel caster dolly that can be removed and cleaned
  • Utilize quick disconnects to attach the mop, sprayer and fogger
  • User-friendly and simplify cleaning and disinfecting procedures
  • Incorporate spray, mop and fog capabilities all in one unit
Certifications
CE marked, ASME rated
Specifications
Product Type Mop
For Use With (Equipment) Core2Clean System
Length (English) 7 in.
Length (Metric) 178 mm