Learn More
VAI Core2Clean System™ Basic Component
Catalog No. 19546987 Shop All VAI Products
Change view
Click to view available options
Volume (English):
2 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-546-987 2 gal.
19-546-988 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-546-987 Supplier VAI Supplier No. C2C102P
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Core2Clean System™ Basic Components are designed to address the application of cleaning and disinfecting agents to the surface in a meaningful and manageable methodology.
  • Made of stainless steel and can be steam sterilized to eliminate the concerns for introduction of viable contamination to the controlled environment from the components themselves.
  • Eliminate concerns for changing of dirtied cleaning solution
  • Incorporate a trigger activated dispensing mop, a sprayer and a fogger all in one unit
  • Present a sufficient wetting of the surface unavailable from other mopping sytems
  • Can reduce cleaning time by 50% and is easily moved from area to area by a 4-wheel caster dolly that can be removed and cleaned
  • Utilize quick disconnects to attach the mop, sprayer and fogger
  • User-friendly and simplify cleaning and disinfecting procedures
  • Incorporate spray, mop and fog capabilities all in one unit
Certifications
CE marked, ASME rated
Specifications
Product Type Spray Tank
For Use With (Equipment) Core2Clean System
Material Stainless Steel
Volume (English) 2 gal.
Volume (Metric) 7.57 L