Learn More
VAI Core2Clean Plus Systems
Catalog No. 19546795 Shop All VAI Products
Change view
Click to view available options
Product Type:
Floor Cover
Mop Frame
Mop Head
Length (English):
7 in.
12 in.
Sterility:
Non-sterile
Sterile
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Length (English) Sterility
19-546-795 Mop Frame 12 in. Non-sterile
19-546-790 Mop Head 12 in. Sterile
19-546-792 Floor Cover 7 in. Sterile
19-546-793 Floor Cover 12 in. Non-sterile
19-546-794 Floor Cover 12 in. Sterile
19-546-791 Floor Cover 7 in. Non-sterile
19-546-803 Mop Head 7 in. Sterile
19-546-802 Mop Head 7 in. Non-sterile
19-546-804 Mop Head 12 in. Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-546-795 Supplier VAI Supplier No. C2C100212
Add to Cart
Add to Cart
Request a Quote
The Core2Clean™ Plus Systems were created by Veltek Associates, Inc., to provide specialized cleaning equipment for use in controlled environments.
  • Core2Clean Plus systems can be configured to operate as three different systems: a sprayer, a wet mop, or a fogger
  • Design of the Core2Clean Plus system allows for repeatability between operators and areas, making it easier for companies to write SOP’s for different class areas
  • Veltek provides an Operator’s Manual with instructions for all three system uses, and videos of select processes
Specifications
Product Type Mop Frame
Head Type Bayonet
Length (English) 12 in.
Sterility Non-sterile
For Use With (Equipment) Core2Clean VertiKlean System
For Use With (Application) For General Cleaning and Wipe down of Walls and Ceilings