Learn More
VAI DECON-CYCLE™ II Germicidal Detergent
Catalog No. 19547025 Shop All VAI Products
Change view
Click to view available options
Volume (English):
16 oz.
Catalog No. Volume (English)
19-547-025 16 oz.
1 options
This item is not returnable. View return policy
Catalog No. 19-547-025 Supplier VAI Supplier No. DCY0616Z01
This item is not returnable. View return policy
Includes
Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report
Phosphate-free
A low pH, dilutable, hospital detergent effective in 10 minutes at 20°C in hard water up to 400ppm (calculated as CaCO3) in the presence of 5% blood serum. Recommended for use in pharmaceutical, biotechnology, medical device manufacturing, and in healthcare institutions to control cross-contamination hazards.
  • Multi-phenolic formula designed to clean, disinfect, and deodorize any washable inanimate surface
  • Filtered at 0.2μm and gamma irradiated
  • Strong enough to kill a broad spectrum of pathogenic bacteria, yet mild enough to have no harmful effect on disinfected surfaces
  • Completely traceable and validated for sterility and shelf life
Order Info
Double-bag packaging.
Compliance
Completely tested according to current USP compendium.
Specifications
Sterility Sterile
For Use With (Application) Kills broad spectrum of pathogenic Bacteria, pharmaceutical, biotechnology, medical device and health care industries
Volume (English) 16 oz.