Learn More
VAI DECON-GLASS Cleaner
Catalog No. 19546837 Shop All VAI Products
Change view
Click to view available options
Volume (English):
16 oz.
Catalog No. Volume (English)
19-546-837 16 oz.
1 options
Catalog No. 19-546-837 Supplier VAI Supplier No. DG0316Z
Add to Cart
Add to Cart
DECON-GLASS Cleaner was developed for use in cleaning rotation cycles to remove residue left behind from disinfecting agents on glass or Plexiglas surfaces. Sterile and formulated with Water For Injection, DECON-GLASS is designed for use pharmaceutical and biotechnology operations.
  • A glass and plexiglass cleaner that is processed to comply with the standards required by pharmaceutical, biotechnology, medical device, and healthcare industries.
Specifications
For Use With (Application) Used for the cleaning of Glass, Plexiglass, surfaces
Volume (English) 16 oz.
Volume (Metric) 473 mL