Learn More
VAI DECON-QUAT 100 Quaternary Ammonium Solution
Catalog No. 19546850 Shop All VAI Products
Change view
Click to view available options
Volume (English):
8 oz.
16 oz.
1 gal.
Sterility:
Non-sterile
Sterile
Format:
Concentrate
SimpleMix
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Sterility Format
19-546-850 1 gal. Non-sterile Concentrate
19-546-851 1 gal. Sterile Concentrate
19-547-054 1 gal. Sterile SimpleMix
19-547-055 1 gal. Non-sterile SimpleMix
19-547-056 16 oz. Sterile SimpleMix
19-546-853 8 oz. Sterile Concentrate
19-547-057 16 oz. Non-sterile SimpleMix
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-546-850 Supplier VAI Supplier No. DQ10001
Add to Cart
Add to Cart
DECON-QUAT 100 Quaternary Ammonium Solution for use on hard, nonporous environmental surfaces in medical and nonmedical facilities.
  • Effective as a one-step disinfectant, virucide, deodorizer, sanitizer and fungicide
  • For use on stainless steel, porcelain, glass and chrome
  • Filled in ISO 5 (Grade A/B, Class 100)
  • Filtered at 0.2 micron
  • Completely validated for sterility and shelf life
  • Hard, inanimate surfaces in filling and gowning rooms, general manufacturing areas, machinery tables, counter tops, and laminar flow benches, floors and wall
Specifications
Sterility Non-sterile
Certifications/Compliance USP, ISO
Composition Active ingredients of 5% Alkyl (60% C14, 30% C16, 5% C12, 5% C18) Dimethyl Benzyl Ammonium Chloride and 5% Akly (68% C12, 32% C14) Dimethyl Ethylbenzyl Ammonium Chloride and 90% inert ingredients
Filtered Filtered
Format Concentrate
For Use With (Application) Used as a disinfectant on inanimate, hard, non porous environmental surfaces such as walls, ceilings, floors, countertops, general manufacturing areas
Volume (English) 1 gal.
Volume (Metric) 3.8 L