Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI DECON-SPORE 200 Plus Disinfectant Solution

Catalog No. 19547294 Shop All VAI Products
Change view
Click to view available options
Volume (English):
0.8 oz.
13 oz.
16 oz.
1 gal.
Format:
Concentrate
Concentrate (Unit Dose)
Concentrate (Unit Dose, Sporicidal Dose with 16 oz. of water)
SimpleMix (3% Disinfectant/Sporicidal Dose)
SimpleMix (Disinfectant Dose)
SimpleMix (Sporicidal Dose)
Sterility:
Non-sterile
Sterile
Packaging:
Attached Trigger Spray
Bottle
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Format Sterility Packaging
19-547-294 16 oz. SimpleMix (Disinfectant Dose) Non-sterile Attached Trigger Spray
19-547-290 0.8 oz. Concentrate (Unit Dose, Sporicidal Dose with 16 oz. of water) Sterile Bottle
19-546-501 13 oz. Concentrate (Unit Dose) Sterile Bottle
19-547-270 16 oz. SimpleMix (Disinfectant Dose) Sterile Attached Trigger Spray
19-547-069 16 oz. SimpleMix (Sporicidal Dose) Sterile Attached Trigger Spray
19-546-591 16 oz. SimpleMix (3% Disinfectant/Sporicidal Dose) Sterile Attached Trigger Spray
19-547-060 1 gal. Concentrate Non-sterile Bottle
19-547-061 1 gal. Concentrate Sterile Bottle
19-547-291 1 gal. SimpleMix (Disinfectant Dose) Non-sterile Bottle
19-547-065 1 gal. SimpleMix (Disinfectant Dose) Sterile Bottle
19-547-292 1 gal. SimpleMix (Sporicidal Dose) Non-sterile Bottle
19-547-066 1 gal. SimpleMix (Sporicidal Dose) Sterile Bottle
19-546-592 1 gal. SimpleMix (3% Disinfectant/Sporicidal Dose) Sterile Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 19-547-294 Supplier VAI Supplier No. DS2000716Z02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Lot-specific Certificate of Analysis, LAL testing analysis and sterility report

DECON-SPORE 200 Plus Disinfectant Solution is designed for the sterilization of manufacturing, packaging and filling equipment in aseptic processes.

  • Decon-Spore 200 Plus is a Sterilant-Virucide-Disinfectant-Sanitizer-Booster for alkaline detergents
  • Effective sanitizer on inanimate, non-porous, non-food contact surfaces
  • Sterile pharmaceutical cleanroom formula
  • Phosphate free
  • Peracetic acid and hydrogen peroxide solution filtered at 0.2 μm
  • Disinfects as it cleans in one operation
  • Broad-spectrum hard surface disinfectant effective against influenza A viruses
  • Filled in ISO 5 (Grade A/B, former Class 100)
  • Double-bagged packaged
  • Available in several container types (unit dose, concentrate and SimpleMix) and sizes both sterile and nonsterile
  • Completely traceable and validated for sterility and shelf life

Simple Mix System

  • Ready to use package
  • Holds concentrate in small chamber and WFI quality water in larger chamber
  • Simply press top of small chamber to release concentrate into water and agitate to mix
  • Eliminates risk of operator error when measuring appropriate amount of concentrate and gives fresh solution every time

Order Info

Suggested minimum surface exposure time for 5% dilution based on the temperature of the solution: 20°C for 6 hours, 50°C for 20 minutes, or 80°C for 5 minutes (68°, 122° or 176°F). In the presence of 5% blood serum and soap film on a nonporous surface, suggested surface exposure time of 10 minutes at 20°C.

Specifications

Sterility Non-sterile
Certifications/Compliance USP, AOAC, ISO
Composition Peracetic Acid and Hydrogen Peroxide solution
Concentration 5% for sterilization, 0.3% for disinfection
Filtered Filtered
Format SimpleMix (Disinfectant Dose)
For Use With (Application) Designed for the sterilization of manufacturing, packaging and filling equipment in aseptic processes, to disinfect floors, walls and other hard non porous surfaces
Includes Lot specific certificate of analysis and certificate of sterility, Attached Trigger
Packaging Attached Trigger Spray
Volume (English) 16 oz.
Volume (Metric) 473 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.