Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI Easy2Gown™ Mitcool 1700 Cleanroom Hoods
GREENER_CHOICE

Catalog No. 19547262 Shop All VAI Products
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-547-262 Large/X-Large
19-546-428 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-547-262 Supplier VAI Supplier No. 1700PHUCS17035
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VAI Easy2Gown™ Mitcool 1700 Cleanroom Hoods are single-use PPE with excellent chemical resistance

  • Folded in an inside-out fashion that essentially elimates the individual from contaminating the outside of the garment
  • Packaged to prevent operator contamination of the outside of the garment
  • Bonded polypropylene with laminate
  • Piped seams with reinforced stitching
  • Excellent chemical resistance

Specifications

Product Type Cleanroom Hood
Size Large/X-Large
Material Polypropylene Laminate
Color White
Disposable Single-use
Seam Type Piped, Stitched
Sterility Sterile
Quantity 100/Cs.
Weight (English) 2.5 oz./sq. yd
Includes Shipped with Lot Specific Documentation
For Use With (Application) Personal Protective Equipment (PPE)
Resistance Chemicals
Hood Type Eye Openings
Cleanroom Class Class 100-1000 (ISO 5-6)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.