Learn More
VAI PF-2 Cleanroom Face Masks
Catalog No. 19546452 Shop All VAI Products
Change view
Click to view available options
Construction:
2-ply
Catalog No. Construction
19-546-452 2-ply
1 options
This item is not returnable. View return policy
Catalog No. 19-546-452 Supplier VAI Supplier No. PF2SMT4S2021
Add to Cart
Add to Cart
This item is not returnable. View return policy
Comfortable material PF-2 face masks for use in pharmaceutical, biotechnology, medical device industries, or any other cleanroom operation.
  • Made from rayon laminated with an acrylic binder for low particulate and shredding characteristics
  • 99% filtration measured at .045 microns
  • Eliminates fogging of goggles
  • Provides breathability
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Ties
Material Rayon
Size Standard
Sterility Sterile
Style Pleated
Construction 2-ply
Disposable Single-use
Filter Efficiency 99%
Includes Four Ties
Model PF-2SM-T-4-S-2021
Packaging Quantity 500/Pk.
Packaging Type Bulk Packaged
Series PF-2