Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI SMA™ Microportable Viable Air Sampler

Catalog No. 19546935 Shop All VAI Products
Change view
Click to view available options
Flow Rate:
28.3, 141.5 L/min.
28.3, 56.6, and 141.5 L/min.
Weight (Metric):
2.7 kg
3.3 kg
4.54 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Weight (Metric)
19-546-935 28.3, 141.5 L/min. 2.7 kg
19-546-937 28.3, 141.5 L/min. 3.3 kg
19-546-936 28.3, 56.6, and 141.5 L/min. 4.54 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-546-935 Supplier VAI Supplier No. SMAP19103
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Includes

Veltek battery charger

The SMA™ Microportable Viable Air Sampler provides consistent environmental monitoring for viable particles.

Battery powered air samplers are intended for use in pharmaceutical clean rooms. The portables draw a measured volume of room air over a sampling plate placed in locations that are critical for environmental quality control sample points.
  • Stainless-steel casing and soft touch controls
  • Rechargeable battery, 8 hours continuous operation
  • Factory-calibrated
  • Plates used are TSA agar type

Warranty and Services

1 Year

Specifications

Material Delrin Top and Bottom, 316L Stainless Steel Housing
Weight (English) 6 lb.
Product Type Microportable Air Sampler
Flow Rate 28.3, 141.5 L/min.
Weight (Metric) 2.7 kg
Overall Height (English) 10 in.
Overall Height (Metric) 25.4 cm
Overall Width (English) 6 in.
Overall Width (Metric) 15.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.