Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI STER-AHOL™ WFI Decontaminant Solution

Catalog No. 19546862 Shop All VAI Products
Change view
Click to view available options
Volume (English):
11 oz.
16 oz.
1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-546-862 16 oz.
19-546-859 1 gal.
19-546-861 11 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-546-862 Supplier VAI Supplier No. DSTERWFITR04
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

STER-AHOL™ WFI Decontaminant Solution disinfects and sanitizes cleanrooms and controlled areas.

Disinfects and sanitizes cleanrooms and controlled areas

  • Made from denatured alcohol formulated to 70% with a small percentage of methyl alcohol and water for injection
  • Assayed according to current USP compendium
  • Filled in ISO 5 (Grade A/B, Class 100)
  • Filtered at 0.2 micron
  • Sterile
  • Nonfood contact, hard nonporous and inanimate surfaces; aseptic filling and gowning rooms; general manufacturing areas and laboratories; machinery, tools, tables, counters, laminar flow benches, floors, walls and carts; gloves

Compliance

EPA registered

Specifications

Sterility Sterile
Certifications/Compliance USP EPA, ISO
Composition Denatured Ethanol made with USP Water for injection, small percentage of Methyl Alcohol and Isopropyl Alcohol
Concentration 70%
Filtered Filtered
For Use With (Application) Disinfectants and sanitizes cleanrooms and controlled areas such as those used in health care institutions, biopharmaceutical, pharmaceutical, medical device and diagnostic manufacturing facilities
Includes Certificate of analysis, certificate of irradiation, LAL and certificate of sterility
Volume (English) 16 oz.
Volume (Metric) 473 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.