Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI STERI-BUFFER™ Phosphate Buffer

Catalog No. 19546921 Shop All VAI Products
Change view
Click to view available options
Volume (Metric):
99 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-546-921 99 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-546-921 Supplier VAI Supplier No. SB10099
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Certificate of Analysis and Certificate of Sterility

STERI-BUFFER™ Phosphate Buffer for disinfectant preparation and dilution.

  • Buffered to a pH of 7.2 ± 0.2
  • Made with USP water for injection filtered at 0.2 μm
  • Filled in easy open-and-close wide-mouth bottles that are scaled on four sides for easy measurement
  • Terminally sterilized through a validated cycle at a SAL level of 10-6
  • Sealed with a no-tamper strip that is broken once the bottle is opened
  • Filled in a ISO 5 cleanroom
  • Validated for sterility

Order Info

Shelf life of two years. Not for parenteral administration.

Compliance

Lot sterility tested per current USP compendium.

Specifications

Product Type Phosphate Buffer
pH 7.2
Sterility Sterile
For Use With (Application) For disinfectant preparation and dilution
Includes Certificate of Analysis, certificate of Sterility
Quantity 72 x 99 mL
Volume (Metric) 99 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.