Learn More
USP Purified Steri-Water, VAI
Catalog No. 19547288 Shop All VAI Products
Change view
Click to view available options
Packaging:
Bottle
Bottle with Unattached Trigger
Gallon Drum, Unattached Faucet
Pail
Volume (English):
16 oz.
1 gal.
2 gal.
5 gal.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Volume (English)
19-547-288 Pail -
19-546-942 Bottle with Unattached Trigger 16 oz.
19-546-941 Bottle 1 gal.
19-546-943 Bottle 2 gal.
19-546-944 Gallon Drum, Unattached Faucet 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-547-288 Supplier VAI Supplier No. STWA8L
Add to Cart
Add to Cart
This item is not returnable. View return policy
USP Purified Steri-Water designed for use in the pharmaceutical, biotechnology, and medical device industries for chemical formulations, cleaning, rinsing and many other applications.
  • Specifically formulated as a sterile pharmaceutical cleanroom formula
  • Lubricates for continuous and effortless manufacturing
  • Designed for all washable environmental surfaces
  • Filled in an ISO 5 (Grade A/B, Former Class 100) operation
  • Filtered at 0.2 microns
  • Components are air washed with 0.2 micron filtered air before assembly
Specifications
Sterility Sterile
Certifications/Compliance Lot specific Certificate of Analysis, Certificate of Sterility, and Certificate of Irradiation
Filtered Filtered
Packaging Pail
Volume (Metric) 8 L